Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Methoxycoumarin 98.0+%, TCI America™
SDP

Catalog No. M172325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M172325G 25 g
M17235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M172325G Supplier TCI America Supplier No. M172325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 531-59-9
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.171
MDL Number MFCD00006876
InChI Key LIIALPBMIOVAHH-UHFFFAOYSA-N
Synonym 7-methoxycoumarin, herniarin, ayapanin, methylumbelliferone, 7-methoxy-2h-chromen-2-one, herniarine, 7-methoxy-2h-1-benzopyran-2-one, 2h-1-benzopyran-2-one, 7-methoxy, coumarin, 7-methoxy, methoxycourmarin, 7
PubChem CID 10748
ChEBI CHEBI:5679
IUPAC Name 7-methoxychromen-2-one
SMILES COC1=CC2=C(C=C1)C=CC(=O)O2

Specifications

Melting Point 118°C
Color White-Yellow
Quantity 25 g
Formula Weight 176.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Methoxycoumarin
RTECSNumber DJ3100380
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.