Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Methoxyindole, 97%

Catalog No. p-7055240
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6376703 1 g
AAH6376706 5 g
AAH6376714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6376703 Supplier Thermo Scientific Chemicals Supplier No. H6376703
Only null left
Add to Cart

CAS: 3189-22-8 | C9H9NO | 147.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3189-22-8
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00047203
InChI Key FSOPPXYMWZOKRM-UHFFFAOYSA-N
Synonym 7-methoxyindole, 7-methoxy indole, 1h-indole, 7-methoxy, indole, 7-methoxy, methoxyindole 7-, unii-9r05qk9rp4, 7-methoxy-indole, pubchem7434, acmc-209hpi
PubChem CID 76660
IUPAC Name 7-methoxy-1H-indole
SMILES COC1=CC=CC2=C1NC=C2

Specifications

Density 1.126 g/mL
Boiling Point 108°C to 110°C (0.3 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.612
Quantity 1 g
Formula Weight 147.17
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 7-Methoxyindole
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-690-1
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.