Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene 95.0+%, TCI America™
SDP

Catalog No. M14431G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14431G 1 g
M14435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14431G Supplier TCI America Supplier No. M14431G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 84030-20-6
Molecular Formula C8H15N3
Molecular Weight (g/mol) 153.229
MDL Number MFCD00043004
InChI Key OEBXWWBYZJNKRK-UHFFFAOYSA-N
Synonym 1,3,4,6,7,8-Hexahydro-1-methyl-2H-pyrimido[1,2-a]pyrimidine, MTBD
PubChem CID 123583
IUPAC Name 1-methyl-2,3,4,6,7,8-hexahydropyrimido[1,2-a]pyrimidine
SMILES CN1CCCN2C1=NCCC2

Specifications

Boiling Point 79°C
Quantity 1 g
UN Number 3267
Formula Weight 153.23
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.