Learn More
Cayman Chemical LTyrosInsodIum salt hydra 100g
Supplier: Cayman Chemical 36333100 G
An AChE inhibitor induces LTP in rat hippocampal slices at 10 UM prevents scopolamine-induced memory deficits in the passive avoidance test in rats from 0 03-1 mg/kg
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.