Learn More
NEXSCREEN LLC 7- Panel Oral Swab Drug Screening Device
Supplier:  NEXSCREEN LLC QODOA10106EUOI
Encompass_Preferred
AMP50, COC20, MET50, OPI40, PCP10, THC40- For employment & insurance use only.
Catalog No. NC2002057
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.