Learn More
Purdue Fredick Company 7 Series® Upper Arm Blood Pressure Monitor
Supplier:  Purdue Fredick Company BP7360
Encompass_Preferred
-Screen for AFib with every measurement; only one measurement is needed for AFib screening-Clinically validated-120 memory total; 2 users x 60 stored readings per user-Large, easy-to-read display and symbols-Date & time stamp on display-Guest mode-Hypertension indicator-OMRON connect app compatible-Pre-formed ComFit cuff (9” – 17”)
Catalog No. NC3596188