Learn More
Stoelting™ 700 Series Microliter Syringe
SDP
Catalog No. 10000731 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Bevel tip
Bevel tip, 6/Pk.
Bevel tip, Removable needle
Bevel tip, Removable needle, 6/Pk
Blunt tip
Luer tip
Luer tip, Needle not included
Size:
10 μL Syringe
10 μL Syringe, 26 gauge.
10 μL Syringes, 26 gauge.
100 μL Syringe
100 μL Syringe, 22 gauge.
25 μL Syringe
25 μL Syringe, 22 gauge.
250 μL Syringe
250 μL Syringe, 22 gauge.
5 μL Syringe, 24 gauge.
5 μL Syringe, 26 gauge.
50 μL Syringe
50 μL Syringe, 22 gauge.
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Size
10-000-731 Luer tip, Needle not included 10 μL Syringe
10-000-713 Blunt tip 10 μL Syringe, 26 gauge.
10-000-715 Blunt tip 50 μL Syringe, 22 gauge.
10-000-716 Blunt tip 100 μL Syringe, 22 gauge.
10-000-717 Bevel tip 5 μL Syringe, 26 gauge.
10-000-718 Bevel tip 10 μL Syringe, 26 gauge.
10-000-719 Bevel tip, 6/Pk. 10 μL Syringes, 26 gauge.
10-000-720 Bevel tip 25 μL Syringe, 22 gauge.
10-000-721 Bevel tip 50 μL Syringe, 22 gauge.
10-000-722 Bevel tip 100 μL Syringe, 22 gauge.
10-000-725 Bevel tip, Removable needle 5 μL Syringe, 26 gauge.
10-000-726 Bevel tip, Removable needle 10 μL Syringe, 26 gauge.
10-000-727 Bevel tip, Removable needle, 6/Pk 10 μL Syringes, 26 gauge.
10-000-728 Bevel tip, Removable needle 25 μL Syringe, 22 gauge.
10-000-732 Luer tip 25 μL Syringe
10-000-733 Luer tip 50 μL Syringe
10-000-734 Luer tip 100 μL Syringe
10-000-735 Luer tip 250 μL Syringe
10-000-737 Bevel tip 250 μL Syringe, 22 gauge.
10-000-919 Blunt tip 5 μL Syringe, 24 gauge.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Catalog No. 10-000-731 Supplier Stoelting™ Supplier No. 51112
Add to Cart
Add to Cart
Stoelting™ Microliter Syringe offers a variety of different terminations and needle options
This original hand-fitted Hamilton syringe is ideal for dispensing volumes from 0.5 μL up to 500 μL.
Specifications
For Use With (Application) Neuroscience
Size 10 μL Syringe
Model Luer tip, Needle not included