Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 7000 Series Copolymer Microsphere Suspensions

Catalog No. 09980354
Change view
Click to view available options
Diameter (Metric):
3.2 μm
6 μm
7.9 μm
11 μm
12 μm
17 μm
19 μm
25 μm
45 μm
55 μm
71 μm
90 μm
97 μm
134 μm
222 μm
Quantity:
15 mL
100 mL
1000 mL
30 product options available for selection
Product selection table with 30 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Quantity
09-980-354 6 μm 100 mL
09-980-350 3.2 μm 15 mL
09-980-351 3.2 μm 100 mL
09-980-352 3.2 μm 1000 mL
09-980-353 6 μm 15 mL
09-980-355 6 μm 1000 mL
09-980-356 7.9 μm 15 mL
09-980-357 7.9 μm 100 mL
09-980-359 12 μm 15 mL
09-980-360 11 μm 100 mL
09-980-361 11 μm 1000 mL
09-980-362 17 μm 15 mL
09-980-363 17 μm 100 mL
09-980-364 17 μm 1000 mL
09-980-365 19 μm 15 mL
09-980-366 19 μm 100 mL
09-980-368 25 μm 15 mL
09-980-371 45 μm 15 mL
09-980-372 45 μm 100 mL
09-980-374 55 μm 15 mL
09-980-375 55 μm 100 mL
09-980-376 55 μm 1000 mL
09-980-377 71 μm 15 mL
09-980-380 90 μm 15 mL
09-980-381 90 μm 100 mL
09-980-383 97 μm 15 mL
09-980-384 97 μm 100 mL
09-980-386 134 μm 15 mL
09-980-387 134 μm 100 mL
09-980-389 222 μm 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 30 options available.
30 options
Catalog No. 09-980-354 Supplier Thermo Scientific™ Supplier No. 7505B
Add to Cart
Add to Cart

Meet the need for particulate materials with a variety of particle sizes and properties with Thermo Scientific™ 7000 Series Copolymer Microsphere Suspensions.

Meet the need for particulate materials with a variety of particle sizes and properties with Thermo Scientific™ 7000 Series Copolymer Microsphere Suspensions, useful as model systems for fluid mechanics experiments and as challenge particles for large pore filtration systems.

Suspensions of large copolymer microspheres are composed of polystyrene polymer, cross-linked with 4 to 8% divinylbenzene (DVB). The particles are chemically inert and can be washed with alcohol and vacuum or air dried. The polymer density is 1.05g/cm3 and the index of refraction is 1.59 at 589nm. They are packaged as aqueous suspensions at 10% solids by weight.

Ordering Information:

15mL, 100mL and 1000mL bottle sizes available. 15mL (“A” Part No. bottles), 100mL (“B” bottles) and 1000mL (“C” bottles) are packaged to order; example (Part No. 7503B and 7503C). Please allow 10 working days when ordering these fill volumes.

Includes:

Material Safety Data Sheet (MSDS) and general product handling insert sheet

Recommended for:

High concentration polymer particles > 3μm used in filter evaluation and testing, fluid mechanics research, dispersion studies and many other research and development projects. Not intended for use in instrument calibration or diagnostic reagents because they lack the exacting specifications needed for those applications. Also useful as experimental particles for acoustical and optical analytical systems.

TRUSTED_SUSTAINABILITY

Specifications

Additive Contains trace amount of surfactant to inhibit agglomeration and promote stability
Concentration 10%
Content And Storage Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.
Description Particles useful as model systems for fluid mechanics experiments and as challenge particles for large pore filtration systems.
Color White or Colorless
For Use With (Application) Fluid Mechanics, Large Pore Filtration
Material Polystyrene Divinylbenzene (PS-DVB)
Product Type Copolymer Microsphere Suspensions
Includes General product handling insert sheet
Packaging 100 mL
Particle Density 1.05g/cm3
Quantity 100 mL
Refractive Index 1.59 at 589nm (25°C)
Stability Product stable for a minimum of 24 months
Diameter (Metric) 6 μm
Show More Show Less

Not intended for use in instrument calibration or diagnostic reagents.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.