Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Silicomolybdic Acid Solution

Catalog No. 70834825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
125 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-834-825G 25 g
70-834-8125G 125 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 70-834-825G Supplier Sigma Aldrich Supplier No. 70834825G
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for:Use in electrostatic synthesis of compounds comprising ultrathin layers of silicododecamolybdate anions and poly(allyl ammonium) cationsPreparation of charge transfer compounds with Keggin anions and 2,2'sa-biquinolinium

Specifications

CAS 11089-20-6
Density 1.156 g/mL at 25°C
Refractive Index n20/D 1.376
Linear Formula H4SiO4 -+ 12MoO3
MDL Number MFCD00054151
Quantity 25 g
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.