Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Ethyl-1-Methylpyrrolidinium Bis(Trifluoromethylsulfonyl)Imide

Catalog No. 7131555G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-315-55G 5 g
71-315-550G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 71-315-55G Supplier Sigma Aldrich Supplier No. 7131555G
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Ethyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide is a 1-alkyl-1-methylppiperidinium-based ionic liquid. It is being investigated for its potential use as a battery electrolyte.1-ethyl-1-methylpyrrolidinium bis(trifluoromethanesulfonyl) imide in combination with 1-propyl-3-methylimidazolium iodide (PMII) forms a solid-state electrolyte with long-term stability, which can be used in dye-sensitized solar cells (DSSCs).

Specifications

CAS 223436-99-5
Molecular Formula C9H16F6N2O4S2
Linear Formula C9H16F6N2O4S2
MDL Number MFCD09038893
Quantity 5 g
Molecular Weight (g/mol) 394.35 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.