Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-(+)-2'-Amino-1,1'-Binaphthalen-2-Ol

Catalog No. 713708500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-370-8500MG 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 71-370-8500MG Supplier Sigma Aldrich Supplier No. 713708500MG
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(R)-(+)-2'-Amino-1,1'-binaphthalen-2-ol is a non-symmetrically substituted 1,1'-binaphthalene ligand.(R)-(+)-2'sa-Amino-1,1'sa-binaphthalen-2-ol may be used as a catalyst in the asymmetric PTC (phase-transfer catalysis) synthesis of +a-amino acids. (R)-NOBIN reacts with trans-azobenzene-4,4'sa-dicarbonyl chloride to form poly(ester-amide)s, which shows backbone light-regulated chiroptical response.

Specifications

CAS 1097192-04-5
MDL Number MFCD04975412
Quantity 25 g
Synonym 2,2,6,6-Tetramethyl-1-piperinyloxy, free radical on silica gel; SiliaCAT(R) TEMPO
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.