Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich O,O'-Bis[2-(N-Succinimidyl-Succinylamino)Ethyl]Polyethylene Glycol

Catalog No. 713783500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-378-3500MG 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-378-3500MG Supplier Sigma Aldrich Supplier No. 713783500MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

O,O'-Bis[2-(N-Succinimidyl-succinylamino)ethyl]polyethylene glycol (NHS-PEG-NHS) is a cross-linking reagent.O,O'sa-Bis[2-(N-Succinimidyl-succinylamino)ethyl]polyethylene glycol (NHS-PEG-NHS) is suitable reagent used in the covalent conjugation of peanut agglutinin and anticarcinoembryonic antigen antibodies (+aCEA) tumor-targeting ligands to near-infrared (NIR)fluorescent superparamagnetic iron oxide (IO)- human serum albumin (HSA) nanoparticles. It is suitable reagent used in the synthesis of novel series of mesoporous silica nanoparticles with different pore sizes, useful for high loading and controlled release of short oligonucleotides.

Specifications

Melting Point 82°C to 86°C
Molecular Formula C13H18BBrO2
Linear Formula C13H18BBrO2
MDL Number MFCD01632210
Quantity 1 g
Synonym 2-(2-(Bromomethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol) 297 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.