Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ 719B Chemical Resistant Unsupported Nitrile Gloves

Catalog No. 17100806 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
Small
Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-100-806 Small
17-100-808 Large
17-100-810 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-100-806 Supplier SHOWA™ Supplier No. 719B07
Add to Cart
Add to Cart

Chemical-resistant unsupported nitrile, 13 in., 11-mil, blue, raised diamond palm.

  • Protecting against a broad range of hazards, the SHOWA 719B chemical-resistant gloves features an extended gauntlet for forearm protection.
  • The 0.28 mm thick unsupported nitrile provides resistance against chemicals, solvents, oils, and animal fats.
  • The ergonomic design ensures comfort during long periods of wear.
  • Featuring a Diamond grip surface: makes wet work easier and safer.
  • 100% Nitrile
  • Unsupported
  • Unlined
  • Chemical Resistant
  • Water Resistant
  • Abrasion Resistant
  • Disposable
  • ForarmProtection
  • Oil Resistant

Specifications

Applications Laboratory
Color Blue
Finger Thickness (English) 11 mil
Finger Thickness (Metric) 0.025 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 11 mil
Palm Thickness (Metric) 0.025 mm
Product Type Chemical-resistant Gloves
Size Small
Thickness (English) 11 mil
Thickness (Metric) 0.025 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.