Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- Protecting against a broad range of hazards, the SHOWA 719B chemical-resistant gloves features an extended gauntlet for forearm protection.
- The 0.28 mm thick unsupported nitrile provides resistance against chemicals, solvents, oils, and animal fats.
- The ergonomic design ensures comfort during long periods of wear.
- Featuring a Diamond grip surface: makes wet work easier and safer.
- 100% Nitrile
- Unsupported
- Unlined
- Chemical Resistant
- Water Resistant
- Abrasion Resistant
- Disposable
- ForarmProtection
- Oil Resistant
Specifications
Specifications
| Applications | Laboratory |
| Color | Blue |
| Finger Thickness (English) | 11 mil |
| Finger Thickness (Metric) | 0.025 mm |
| Latex Free | Yes |
| Material | Nitrile |
| Palm Thickness (English) | 11 mil |
| Palm Thickness (Metric) | 0.025 mm |
| Product Type | Chemical-resistant Gloves |
| Size | 2X-Large |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?