Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cyanomethyl dodecyl trithiocarbonate

Catalog No. 7230295G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-302-95G 5 g
72-302-91G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 72-302-95G Supplier Sigma Aldrich Supplier No. 7230295G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Need help choosing the correct RAFT Agent? Please consult the RAFT Agent to Monomer compatibility table.Reversible Addition Fragmentation Chain Transfer (RAFT) Polymerization

Specifications

CAS 796045-97-1
Molecular Formula C15H27NS3
Linear Formula C15H27NS3
Quantity 5 g
MDL Number MFCD11045090
Synonym S-Cyanomethyl-S-dodecyltrithiocarbonate
Molecular Weight (g/mol) 317.58
Percent Purity 98% (HPLC)
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.