Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethylmethyl(2-Methoxyethyl)Ammonium Bis(Trifluoromethylsulfonyl)Imide

Catalog No. 7276795G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-767-95G 5 g
72-767-925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 72-767-95G Supplier Sigma Aldrich Supplier No. 7276795G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diethylmethyl(2-methoxyethyl)ammonium bis(trifluoromethylsulfonyl)imide is a room temperature ionic liquid (RTIL), which shows high CO2 solubility.Diethylmethyl(2-methoxyethyl)ammonium bis(trifluoromethylsulfonyl)imide can be used to develop a polymeric gel electrolyte system that shows magnesium ion (Mg2+) mobility.

Specifications

CAS 464927-84-2
Molecular Formula C10H20F6N2O5S2
Linear Formula C10H20F6N2O5S2
MDL Number MFCD06656282
Quantity 5 g
Synonym N,N-Diethyl-N-methyl-N-(2-methoxyethyl)ammonium bis(trifluoromethanesulfonyl)imide
Molecular Weight (g/mol) 426.4 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.