Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Allyl-3-Methylimidazolium Bis(Trifluoromethylsulfonyl)Imide

Catalog No. 72769550G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-769-550G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 72-769-550G Supplier Sigma Aldrich Supplier No. 72769550G
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Allyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide is an ionic liquid that can be prepared by reacting 1-allyl-3-methylimidazolium chloride and lithium bis(trifluoromethylsulfonyl)imide under microwave conditions.1-Allyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide can be used to prepare AMIMTFSI-propylene carbonate based electrolyte, with potential application in lithium-ion batteries. It also finds application in alkanes/alkenes separation process. AMIMTFSI can be used as a surface protector to develop luminescent silicon nanoparticles.

Specifications

CAS 917956-73-1
Molecular Formula C9H11N5
Linear Formula C9H11N5
MDL Number MFCD16875658
Quantity 5 g
Molecular Weight (g/mol) 189.22 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.