Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ 7300 Real Time PCR Systems Spectral Calibration Kit
GREENER_CHOICE

Catalog No. 4349182
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-491-82 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-491-82 Supplier Applied Biosystems™ Supplier No. 4349182
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Easy-to-use, preloaded, 96-well optical reaction plates enable quick and easy calibration of the 7300 Real-Time PCR System.

This easy-to-use kit establishes the pure dye spectra and multicomponent values for FAM™/SYBR™ Green I, VIC™/JOE™, and NED™/TAMRA™/ROX™ dyes on the Applied Biosystems™ 7300 Real-Time PCR System. The Applied Biosystems™ 7300 Real-Time PCR Systems Spectral Calibration Kit I contains one background plate, one region of interest (ROI) calibration plate, and seven spectral calibration plates with seven separate dye standards.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains one background plate, one ROI calibration plate,and seven spectral calibration plates with FAM™⁄SYBR™ Green I, VIC™⁄JOE™, and NED™⁄TAMRA™⁄ROX™ dye standards. Store at -15 to -25°C. Protect from light.
Calibrated Dye FAM, JOE, NED, ROX, TAMRA™, VIC, SYBR™ Green
For Use With (Equipment) 7300 System
Detection Method Fluorescence
Product Type Spectral Calibration Kit
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Dry Ice
Quantity 1 Kit
Calibration Type Spectral Calibration
What type of Real-Time PCR (Sequence Detection System) information is available from Applied Biosystems?

Tutorials demonstrating introductions to Real-Time PCR, guidelines for designing real-time assays using Primer Express software, and various instrument set-ups can be found under Life Technologies University on our website. User bulletins, manuals and Product inserts, protocols can be found as downloadable pdf files on our Technical Resources page.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.