Learn More
Sigma Organic Chemistry Acetonitrile | 1L | 75-05-8 | MFCD00001878 | 99.999% trace metals basis
Supplier: Sigma Organic Chemistry 7334661L

Acetonitrile, 1L
About This Item:
Synonym(s): ACN; Cyanomethane; Ethyl nitrile; Methyl cyanide
Linear Formula: CH3CN
Molecular Weight: 41.05
CAS Number: 75-05-8
MDL Number: MFCD00001878
UNSPSC Code: 26111700
Purity: 99.999% trace metals basis
Boiling Point: 81-82 C (lit.)
Melting Point: -45 C (lit.)
Density: 0.786 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.344 (lit.)
Storage: room temp
EINECS Number: 200-835-2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.