Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Silver nanowires

Catalog No. 73942125ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
73-942-125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 73-942-125ML Supplier Sigma Aldrich Supplier No. 73942125ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Silver nanowires (AgNWs) are 1-D silver based nanostructures which have a diameter of 60 nm and a 10 ++m length. They can be synthesized by the reduction of silver nitrate (AgNO3) in presence of silver bromide (AgBr). They may also be prepared by the arc discharge mechanism of immersed silver electrodes.Silver nanowires are useful in a wide variety of conductive, optical and anti-microbial applications such as touchscreen displays, medical imaging and sterile clothing.

Specifications

CAS 7440-22-4
Density 0.785 g/mL (at 25°C)
Quantity 25 mL
Molecular Formula Ag
Linear Formula Ag
MDL Number MFCD00003397
Synonym Silver nanofibers; Silver nanowhiskers; Silver nanowire
Molecular Weight (g/mol) 107.87
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.