Learn More
Sigma Organic Chemistry 3-aminomethylphenylboronic acid | pinacol ester hydrochloride | 1G | 380430-65-9 | MFCD03410294
Supplier: Sigma Organic Chemistry 7412051G
3-aminomethylphenylboronic acid, pinacol ester hydrochloride, 1G
About This Item:
Linear Formula: C13H21BClNO2
Storage: 2-8C
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.