Learn More
Scott Safety™ 742 Series Air-purifying Respirator Accessory, Cartridges
Description
- Low profile, swept back cartridge design ensures optimal field of vision and allows use of communications
- Available in a full range of NIOSH approved, chemical cartridges or combination cartridges
- Attaches directly to XCEL Halfmask and Promask full facepiece
- Uses Quarter turn adapter when used with AV Series Facepieces
Specifications
Specifications
| Cartridge Type | Particulate, P100 Cartridge |
| Certifications/Compliance | NIOSH |
| Color | Magenta/Magenta |
| Filter Type | Particulate |
| Product Type | Cartridge/Filter Combination |
| For Use With (Application) | Non-IDLH and non-O2 deficient applications |
| For Use With (Equipment) | Scott 742 Series Air Purifying Respirators |
| Quantity | 1 Pair/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.