Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Octadecene

Catalog No. 74740500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-740-500ML 500 mL
74-740-100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 74-740-500ML Supplier Sigma Aldrich Supplier No. 74740500ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Octadecene

Specifications

CAS 112-88-9
Melting Point 14°C to 16°C (lit.)
Density 0.789 g/mL (at 25°C (literature))
Boiling Point 314.4°C ± 5.0°C (760 mmHg), 179°C (15 mmHg), 314.4°C ± 5.0°C (760 mmHg)
Molecular Formula C18H36
Linear Formula CH3(CH2)15 CH=CH2
Refractive Index n20/D 1.444 (literature); n20/D 1.445
Quantity 500 mL
MDL Number MFCD00009003
Molecular Weight (g/mol) 252.48
Percent Purity ≥95.0% (GC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.