Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ 7500 Fast Real-Time PCR System, laptop

Catalog No. 4351106
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 system
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-511-06 1 system
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-511-06 Supplier Applied Biosystems™ Supplier No. 4351106

Item Discontinued This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or visit www.fishersci.com/quantstudio for more information and to contact a Life Science Specialist.

missing translation for 'validMainframeMsg'

Includes

Laptop Computer

Offers maximum performance in the minimum time - fully optimized for Fast cycling, the 7500 Fast delivers high-quality results in as little as 30 minutes.

We supply a Dell™ Notebook computer with the 7500 Fast System.

  • Specially designed Fast block ensures thermal uniformity at top speeds.
  • Faster ramp rates enable rapid results without compromising extension times or assay quality.
  • Fast optical plates ensure excellent precision in 10 to 30μL reaction volumes.
  • Use TaqMan™ Fast Universal PCR Master Mix to run multiple Fast assays on one plate.
  • Finish more runs per day with the excellent performance equivalent to that offered by the 7500 System.

High Resolution Melting Analysis:
The Applied Biosystems™ High Resolution Melting (HRM) Software is the easiest to use melting analysis software enabling real-time PCR melt curve assays to be used more accurately for mutation scanning and genotyping. The HRM Software is available on the 7500 Fast Real-Time PCR System, which delivers precise results in a standard 96-well format.

Use the Applied Biosystems™ High Resolution Melting (HRM) Software to perform more sophisticated melting analysis with an easy to follow workflow and minimal subjective data analysis steps.

  • Shortens analysis time by auto-calling genotypes and automatically omitting the no template controls.
  • Minimizes subjective analysis by automatically grouping unknown variant clusters.
  • No temperature shift required – distinguish between homozygous mutants and wild type more easily.
  • Allows easy data review with customizable multi-plot views, expandable windows and one-click color assignment to highlight curves of interest.
  • Facilitates data presentation with the option to export data and graphs directly to PowerPoint or as JPEG files.

NEW - 7500 Software v2.0:
Now the easy-to-use StepOne software is available for both the 7500 and 7500 Fast systems with the 7500 Software v2.0 upgrade. The 7500 Software v2.0 incorporates your favorite StepOne Software features, such as a variety of plate setup wizards, standard curve dilution and master mix recipe calculators, QC flags, data filters, and email notification when a run is finished. The 7500 Software v2.0 also includes an enhanced Gene Expression Study package and has a variety of new melting curve protocol options, including multiple peak detection, step and hold temperature control, and customizable ramp rates.

The Gene Expression Study package accommodates large studies better than any other instrument software package:

  • Import an unlimited number of Comparative CT (relative quantitation) files to one study.
  • Group samples and view data both by technical replicate group and biological replicate group.
  • Use any gene(s) as an endogenous control, including averaging multiple controls together.
  • Enter known efficiency values to be factored into the RQ results

21 CFR Part 11 Module available:
The SDS v1.4 21CFRp11 Module is a powerful tool for assisting with 21CFRp11 compliance, while still offering the flexibility of user-customizable configuration settings.

  • Individual user log-ins can be added for up to four user groups, each group with designated permission settings.
  • User-customizable permission settings include fourteen system activities, e-signature authority designation, and additional security settings to give you maximum control over your compliance efforts.
  • Audit trails can be enabled or disabled depending on your traceability needs.
  • A selection of e-signatures is available to ease e-signatures into your workflow.

One System, Many Applications:
Applications include gene expression analysis, pathogen quantitation, SNP genotyping, isothermal and +/- assays utilizing internal positive controls. To facilitate many of these applications, Applied Biosystems™ provides preformulated, ready-to-use, quality-tested, TaqMan™ assays for use with the 7500 Fast system. Now you can reduce your assay optimization efforts.

For Research Use Only. Not for use in diagnostics procedures.

TRUSTED_SUSTAINABILITY

Specifications

Format 96-well Plate, 8-well Strips
Thermal Accuracy ±0.5°C
Sample Ramp Rate Fast Mode: ± 2.2°C⁄s, Standard mode: ± 1.6°C⁄s
Sensitivity Down to 1 Copy of Human RNAse P gene
Temperature Range (Metric) 4.0 to 99.9°C
Reaction Volume Range 10–30 μL
Passive Reference Dye ROX (Separate Tube), Other Passive Reference Dye, ROX (Pre-mixed)
Dynamic Range 9 Logs of Linear Dynamic Range
Content And Storage Laptop Computer
Calibrated Dye TAMRA, Cy3, FAM, SYBR Green, Texas Red, JOE, ROX, Cy5, NED, VIC
For Use With (Equipment) 7500 Fast System
Thermal Uniformity ±1°C
Reaction Speed Fast, Standard
Throughput 4 to 5 96-well Plates per day
Run Time 36 min. (Fast), <2 hrs (Standard)
Optics CCD Camera, Halogen Light Source, Five Excitation and Five Emission Filters
Thermal Cycling System Peltier-based System
Dimensions 34 cm (13.39 in.) (W) x 49 cm (19.29 in.) (H) x 45 cm (17.72 in.) (D)
Capacity 12 x 8-tube Strips, 1 x 96-well Plate (Fast)
External Computer Laptop Computer Provided
Precision Distinguish between 5000 to 10,000 genome equivalents (two-fold Copy Number difference) with 99.7% Confidence
Peak Block Ramp Rate 5.5°C⁄s
Max. Heating Rate (Thermal Block) 5.5°C
Quantity 1 system
Weight 34 kg (75 lb.)
Show More Show Less
Why are there no RQ values for all the samples in my study from the 7500 or 7500 Fast Real-Time PCR System software?

The 7500 software requires that the endogenous control be run on every plate within a study. If not, you will only see RQ values for the plate that contains the endogenous control on it. The other plates will have CT values for all the wells, but the software will not calculate RQ values.

If you designed your experiments in this type of layout, you will want to do your analysis in the free DataAssist software instead. This program does not have the endogenous control requirement for every plate. Simply export the study results file from the 7500 software as a *.txt or *.csv file, and use that to create a new study in the DataAssist software. You should now be able to see RQ values for all samples, even those without an endogenous control on the same plate.

Why do I not see any CT or RQ values from my ddCT experiment in the software for the 7500 or 7500 Fast Real-Time PCR System?

If you have the SDS v1.4 or earlier, you will need to open the dd CT plate in a Study first in order to see the CT or RQ values. (If you have v2.0.x this error does not apply.)

1.After the initial run is complete, open, review the plots and save it. Then:
-Choose File then New
-Choose: Assay then ddCT (Relative Quantitation) Study. Set the other parameters as appropriate and click Next.
2.Click on ‘Add Plates...', then browse to where your data file is located and select the plate (or multiple plates).
Click ‘Finish', and you should see your study.

What can I do when I get the error message "Time too short" on the 7500 or 7500 Fast Real-Time PCR System?

You may encounter this error message, which will abort the run before it even starts. If so, go into the software under Setup then Run Method and check how much time has been set for the data collection step. The instrument has a minimum collection time, which is dependent on the number of filters being used. (This is true in both the 1.x and 2.0.x versions of the 7500 software.) Increase the extension time to 30 sec and restart the run. It should now proceed as normal. If the error persists, please contact us at techsupport@thermofisher.com for further assistance.

What can I do if I get an error message about "Block Up Switch" on the 7500 or 7500 Fast Real-Time PCR System?

This error can be caused by the wrong plastics being used in the block, such as the wrong Precision Plate Holder being used in the tray. This can give an error message such as “Block up switch not activating” and will not allow the run to start. Open the drawer, make sure the correct plate/tubes and plate/tube holder are being used and in the correct orientation. Turn off the instrument and open the front cover to make sure the heated cover door is closed. Restart the instrument and retry the run. If the error still persists, please contact us at instrumentservices@thermofisher.com for further assistance.

On the 7500 and 7500 Fast Real-Time PCR Systems, what can I do if I get an error message about "CCD acquisition failure"?

If you see this error message, please check the following:

1. Verify that the USB connection is secure between the instrument and computer.
2. Verify that the computer user power save settings are set to Never for all users (in Windows under Control Panel, Power Options).
3. Is there a screen saver in use? If so, set the screen saver so that it will not come on during a run.
4. Is there antivirus or other network software in use? If so, disconnect from the network while running the instrument.
If the error persists, please contact us at instrumentservices@thermofisher.com for further assistance.

What can I do if the ROI calibration does not pass on the 7500 or 7500 Fast Real-Time PCR System?

Sometime there is an error such that the ROI will not calibrate all 96 wells. If you run into this problem check the following:

- Is the ROI calibration plate old or expired?
- Spin down the plate and visually inspect the wells. Is the volume consistent and correct across all wells? If not, try a new ROI calibration plate.
- Try the manual calibration and increase the exposure time.
- Check the status of the halogen lamp. If it is over 2,000 hours, replace the bulb and try again.

If you have tried all the steps above but the ROI still will not calibrate, please contact us at instrumentservices@thermofisher.com for further assistance.

What can I do if the heated cover is not getting to the set temperature on the 7500 or 7500 Fast Real-Time PCR System?

You may get a message that the heated cover is not reaching the set temperature. If you get this error, or notice this issue on your own, please contact us at instrumentservices@thermofisher.com.

What should I do if I get "Fatal Error 1420" in SDS 2.0.x software on the 7500 or 7500 Fast Real-Time PCR System?

“Fatal Error 1420” is related to the door or heated cover not being fully closed on the instrument itself. We recommend opening the front access door using a small screwdriver, ensuring that all inner components are correctly seated, then closing the door completely. Please then reboot the instrument and attempt to restart your run. Please see page 60 of the Installation and Maintenance Manual for more information: https://tools.thermofisher.com/content/sfs/manuals/cms_041437.pdf

What can I use for data analysis?

Click on the ‘Group By' button and choose ‘Well Position' (Column). Then highlight the plate and print the report.

How can I export or print by Column instead of Row in the instrument software?

Go to ‘View Well Table' from the Amp B117Plot screen.

Can I use the 7500 or 7500 Fast Real-Time PCR System to do protein thermal shift experiments?

Yes. If you have the newer version of the software (v2.0.1 or later), which creates *.eds files, your data will be directly compatible with our Protein Thermal Shift Software (https://www.thermofisher.com/order/catalog/product/4466038?ICID=search-product). If you have the older software (v 1.x), you will have to program the software differently (see below) and analyze the results independently. For more details on the analysis, you can refer to this paper: “The use of differential scanning fluorimetry to detect ligand interactions that promote protein stability.” Nat Protoc 2007;2(9):2212-21.

For an experiment using SDS v1.x, follow the directions below:
1.Create a new experiment, choosing Absolute Quantification assay type. Click ‘Next'.
2.Go to ‘Select Detectors'. If there is no predefined detector for this application, create a new one. Make sure the Reporter Dye is ROX, and Quencher Dye is none.
3.Assign the detector to the plate. Select none for the Passive Reference

How do I set up a touchdown PCR experiment?

In a touchdown PCR experiment, you will either change the temperature or the time of a particular PCR step with every cycle. Most commonly, the annealing temperature is adjusted throughout the experiment, such that the specificity is increased in the early cycles and the efficiency in the later cycles.

In this example, we will set the method to do the following:
1. Go to File then New Experiment then Advanced Setup. Fill out the relevant options as you normally would.
2. Go to the Run Method under the ‘Setup' section and you should see the Graphical View of your thermal profile. Check the box next to ‘Enable AutoDelta'. You should see some grey triangles appear next to the Temperature and Time at every step in the Cycling Stage. (Note: If you want to start the changes at a later cycle, set this here under ‘Starting Cycles'.)
3. A new window called ‘AutoDelta Settings' will open up. Select the appropriate options. In this example we are decreasing the temperature by 0.4 degrees C per cycle, so choose (“-“) and (0.40). Click ‘Save Setting'. You will then see a green triangle show up next to the parameter you changed, in this case next to the 72 degrees C step. Your new method has now been applied

Where are my files saved in the 7500 and 7500 Fast Real-Time PCR System software?

Run files will be saved to a default folder on the connected computer, unless you change it. To find or change the default folder (in SDS v2.0.1 or later), go to Tools then Preferences then Defaults. Here you will see a Data Folder and an Import Folder. The default location is shown. If you want files to be saved to (or open from) a different location, click ‘Browse' and choose the new folder.

Can I collect data at two separate steps for analysis in the 7500 and 7500 Fast Real-Time PCR Systems?

No. The software will use the last data collection step in the cycling stage for all amplification plots and Ct analysis. So even if you were to set two separate steps with “Data Collection On”, you would only be able to view and analysis data from the latter step.

What volumes can be used in my plate for the 7500 or 7500 Fast Real-Time PCR System?

The following volumes are supported for each instrument block:
-7500: 20-100 µL reactions
-7500 Fast: 10-30 µL reactions

Can I use the 7500 or 7500 Fast Real-Time PCR System computer while a run is in progress?

Using the instrument computer when a run is in progress is not recommended, as this poses a risk of corrupting the data.

How do I add a custom dye to the 7500 or 7500 Fast Real-Time PCR System?

The 7500 and 7500 Fast Real-Time PCR Systems can be used to run assays designed with custom dyes (dyes not manufactured by Life Technologies). Custom dyes must fluoresce within the 520-650 nm spectral range measured by the 7500 or 7500 Fast instrument. To use a custom dye, you must first determine what the right concentration of the dye is. You will need to order an oligo with a 5' custom dye but no quencher. Make up a plate with different concentrations of this oligo (approximately 25-3,200 nM) and use the ROI Inspector to assess fluorescence. Choose the concentration that displays the brightest possible signal without saturation in all filters. Once you have found the correct concentration, create a full plate of custom dye at this concentration and perform the custom dye calibration. See Appendix B in the 7500 and 7500 Fast Real-Time PCR System Maintenance Guide (http://tools.thermofisher.com/content/sfs/manuals/cms_077749.pdf) for full details.

What dyes can I use on the 7500 and 7500 Fast Real-Time PCR Systems?

The Applied Biosystems 7500 and 7500 Fast Real-Time PCR Systems use the following dye sets for calibration: Cy3, Cy5, FAM, JOE, NED, ROX, SYBR Green, TAMRA, Texas Red, and VIC dyes. Custom dyes that are read between 520 and 650 nm can also be used, although you will have to calibrate the system first for any new dye.

What are the specifications for the 7500 and 7500 Fast Real-Time PCR Systems?

Instrument Specifications
Block Options (Fixed): 7500: 96-well (standard); 7500F: 96-well (Fast)
Sensitivity: Down to 1 copy
Dynamic Range: 9 logs of linear dynamic range
Calibrated Dyes: FAM, SYBR, VIC, ROX, NED, TAMRA dyes (Cy3, Cy5, and Texas Red dyes - 7500F only)
Detection Method: SYBR dye, primer-probe detection
Resolution: Detect changes as little as 1.5-fold
Reaction Volume Range: 20-100 µL (7500); 10-30 µL (7500F)
Reaction Speed: Fast or standard (7500 has standard mode only)
Optics: Tungsten-halogen lamp, 5 excitation filters, 5 emission filters, CCD Camera
Temperature Range: 4-99.9 degrees C
Run Time: <2 hr (standard mode); ~35 min (Fast mode - 7500F only)
Temperature Accuracy: ±0.25 degrees C (between 35 degrees C and 95 degrees C, after 3 min)
Temperature Uniformity: ±0.5 degrees C (after 30 sec)
Thermal Cycling System: Peltier-based system
Available Applications: Gene expression, genotyping, copy number variation, HRM, protein thermal shift, protein detection, mutation detection, miRNA, presence/absence
Dimensions: 34 cm (W) x 45 cm (D) x 49 cm (H)
Weight: 34 kg (75 lb)
Remote Monitoring: No
On-Board Memory: No
Setup Configurations: PC-controlled only

What do I do if the background calibration fails in the 7500 or 7500 Fast Real-Time PCR System?

Follow the directions in the 7500 and 7500 Fast Real-Time PCR System Maintenance Guide, Chapter 6 (http://tools.thermofisher.com/content/sfs/manuals/cms_077749.pdf), to decontaminate the sample block. You can also watch this helpful video demonstration (https://www.youtube.com/watch?feature=player_embedded&v=dJyolVGhpjk).

When do I need to run the RNaseP calibration in the 7500 or 7500 Fast Real-Time PCR System?

The RNaseP verification plate contains template, master mix, and a TaqMan assay for RNaseP. It is used to verify that the instrument is performing to specifications. If you have reason to suspect there is something wrong with the instrument, if the instrument has been moved, or if you want to rule out a chemistry issue, the RNaseP plate is a good way to test the system. The RNaseP verification plate is a single-use plate.

Can I reuse my calibration plates for the 7500 or 7500 Fast Real-Time PCR System?

The calibration plates can be stored and reused three times for up to 12 months after you first open them, so make sure to return them to their original packaging and return them to -20 degrees C storage until the next use. If needed, you can make your own background plate using deionized water. Please follow the directions in the 7500 and 7500 Fast Real-Time PCR System Maintenance Guide (Appendix C) (http://tools.thermofisher.com/content/sfs/manuals/cms_077749.pdf) for more details.

When do I need to calibrate the 7500 or 7500 Fast Real Time PCR System?

Recommended Maintenance Schedule

Power on/off the computer controlling the instrument: Weekly
Check computer disk space. If necessary, archive or back up your experiment files and instrument settings: Weekly
Background calibration: Every month
Run disk cleanup and disk defragmentation: Every month
Perform an instrument self test: Every month
Pure dye calibrations: Every 6 months
ROI calibration: Every 6 months
Optical calibration: Every 6 months
RNaseP instrument verification: After the instrument has been moved, or as needed to verify instrument performance

Can TET dye be used on the Applied Biosystems real-time PCR instruments?

TET dye is not recommended to be used on the following real-time PCR instrument models: 7000, 7300, 7500, 7500 FAST, StepOne, and StepOnePlus Systems.

Why is the ADD DISSOCIATION CURVE button disabled in the Relative Quantification Plate Assay in the SDS software on the ABI PRISM 7000, Applied Biosystems 7300, 7500, 7500 Fast and the 7900HT Systems?

The older versions of the software did not include this feature for all assay types. For the 7500 and 7500fast instruments, the latest software 2.0.5 allows a melt curve to be added to the PCR experiment. The other instruments/software require a separate program for melt curves. To run a Dissociation Curve after running a Relative Quantification Plate (ddCt) Assay, you will need to open a new assay from File -> New, select DISSOCIATION from the assay pull-down menu and run the Dissociation Curve as a separate assay. This will generate a separate SDS file for the Dissociation Run.

What is the maximum thermal output for the Applied Biosystems 7500 / 7500 Fast Sequence Detection System in BTU/h?

The maximum thermal output for the Applied Biosystems 7500/7500 Fast Sequence Detection System is 3241.5 BTU/h (950W).

What are the ramp rates for the Applied Biosystems 7500 and 7500 Fast Real-Time PCR Instruments?

The sample ramp rates for the 7500 instrument are as follows:

- Standard Mode: 1.6 deg C/sec up and 1.6 deg C/sec down
- 9600 emulation Mode: 0.8 deg C/sec up and 1.6 deg C/sec down. This matches the sample ramp rate achieved for the ABI PRISM 7700 Sequence Detection System.
- Fast Mode: 3.5 deg C/sec up and 3.5 deg C/sec down.

How can I export the amplification plot or gene expression plot from the Applied Biosystems 7300, 7500, and 7500 Fast Real-Time PCR System’s software?

With SDS 1.4 software, right-click on the graph, and choose "Export as JPEG" or "Export to Powerpoint". With SDS 2.0.x (for 7500/7500fast only), you can either click on the save icon above the graph "Save as JPEG Image" or click on the Export drop-down arrow (located on the upper toolbar) and choose "Send to Powerpoint"

Can I move the Applied Biosystems 7300, 7500, or 7500Fast Real-Time PCR System to another room or another building by myself?

Yes, you can move the instrument carefully on your own. It is recommended that you recalibrate the instrument with the pure dye calibration kit(P/N 4349182, 4349180, 4351151, 4360788, or 4362201) and run an RNase P instrument verification plate (P/N 4351979 or 4350584), after the move.

How can I run a standard TaqMan assay reaction on the Applied Biosystems 7500 Fast Real-Time PCR System?

You can run an Applied Biosystems 7500 Fast Real-Time PCR System instrument in Standard mode as long the following set-up points are followed:

1) The reaction volume is between 10 ul-30 ul
2) The 96-well plate is a MicroAmp FAST 96 well reaction plate (P/N 4346907, 4346906 or 4366932)
3) Select the Run Mode to be “Standard 7500 “or “9600 Emulation” (2.0.x software versions only offer Standard or Fast - no 9600 emulation choice).

What are the extensions of files generated by the ABI PRISM 7000 Sequence Detection System, Applied Biosystems 7300, 7500, 7500Fast, 7900HT Fast Real-Time PCR System, StepOnePlus Real-Time PCR System, and ViiA 7 Real-Time PCR Systems?

All run files have an .SDS or .EDS extension. A Relative Quantification study will have an .SDM or .EDM extension and a template will have an .SDT or .EDT extension. The software for StepOnePlus, ViiA 7, and 7500 2.0.x software have .EDS, .EDT, or .EDM extensions; all other software have .SDS, .SDT, or .SDM extensions.

What dyes can be used as a reporter in a multiplex reaction on Applied Biosystems 7500 Real-Time PCR System?

The following dyes can be used as the reporter dye with MGB-NFQ as the quencher on the Applied Biosystems 7500 Real-time PCR System or Applied Biosystems 7500 Fast Real-Time PCR System when using TaqMan Gene Expression Master Mix (recommended for multiplexing) or any other Applied Biosystems TaqMan master mix: FAM, VIC, JOE, NED, TAMRA,CY3, ROX, TEXAS RED, CY5 dyes.

Of these dyes, we recommend a duplex reaction with FAM and VIC or FAM and NED dyes; if you are triplexing, we recommend FAM, VIC, and Cy5 or FAM, VIC, and TAMRA dyes. You must choose dyes that are well separated in emission wavelength. If you are using any probes with a TAMRA dye quencher, you cannot use NED, TAMRA, or CY3 dye as one of the reporter dyes in that multiplex reaction. ROX or TEXAS RED dyes can be used as reporter dyes only when not using an Applied Biosystems Master Mix.

I lost my user manuals for the 7300 and 7500 Fast Real-Time PCR Systems. How can I get an additional copy?

Please contact Technical Support for Real-Time PCR, and provide the serial number of your instrument. We will email you a copy of the user manual upon request.

I am using 7500/7500F Software v1.x. How do I fix the error "Failed to load calibration file C:\Applied Biosystems\7500 System\Calibration\Calibration.sda. Error-100"?

There are two potential ways to fix this error:
1. Re-install the software to get back the default calibration.sda file.

2. Replace the calibration.sda file with the working copy from another computer that has the same 7500 software installed (or if you had previously backed up the software).

Please note a run can be started after replacing the calibration.sda but different calibration audit trail information will be shown in the software (if you are using the 21CFRp11 version of the software).

What are the thermal cycling parameters of the Fast mode for the 7500 Fast Real-Time PCR System?

For the 7500 Fast real time PCR systems, the thermal cycling parameters are defaulted to 50°C/2 min for UNG incubation, 95°C/20 sec for fast polymerase activation, 40 cycles of 95°C/3 secs for the dissociation and 60°C/30 seconds for annealing and extensions.

Can I run slower, standard thermal cycling experiments on a 7500 Fast Real-Time PCR System?

Yes. you can choose either standard or fast ramp on the 7500 Fast System for software versions 2x or greater, and a 9600 emulation thermal cycling mode is available for customers who wish to run assays based on adjusted ramp rate. However, you should be aware that we cannot guarantee identical performance to the same modes run on a standard 7500 system block.

Can I compare the data collected when using Fast thermal cycling mode compared to using standard thermal cycling mode on the 7500 Fast Real-Time PCR System?

Runs can be compared between standard and fast mode, as the data collected are collected in all five filters in the instrument. However, keep in mind that these data may not always show identical results and may have added variability due to the data collection processes and differences in ramp rate between standard and fast mode.

If I were to upgrade my existing 7500 System standard unit to the a 7500 Fast configuration, can I still open my old data files?

Yes, older 7500 or 7500 fast data files are compatible with the current version of the SDS software. However, there is one exception: please note that the SDS v1.3 software will not allow RQ Studies to be created from RQ Plates from different instrument types, different thermal modes (i.e., fast and standard modes) or different numbers of thermal cycles.

Why am I unable to calibrate for any other dye, except for FAM, with my 7500 Fast Real-Time PCR System, laptop (Cat. No. 4351106)?

Most likely you cannot see the option for calibrating with another dye due to the size of the software screen you are viewing. When going through the calibration wizard, make sure to scroll down when on the 'Overview' window to see all options.

Which computer operating systems are compatible with the7500 Fast Real-Time PCR System?

You can use up to Windows XP, Service Pack 3 (SP3) on the computer that is connected to the 7500 Fast Real-Time PCR System instrument. An exception is the SDS v2.0.5 software, which can be used with the Windows 7 operating system for stand-alone data analysis steps.

What is the recommended input template amount for each quantitative reaction on the 7500 Fast Real-Time PCR System?

The recommended input template range is 10-100 ng of gDNA/ cDNA template per reaction.

What is the recommended reaction volume that I can run for using 7500 Fast Real-Time PCR System?

We recommend running a total reaction volume between 10 and 30 ul per well.

To analyze an RQ Study, can I import RQ plates from a combination of Fast and Standard mode runs on a 7500 Fast Real-Time PCR System?

We do not recommend this as the current and previous versions of SDS software will not allow RQ Studies to be created from RQ Plates from different instrument types, different thermal modes/profiles or different numbers of thermal cycles. It is important to reduce variability among RQ plates in an RQ study by importing RQ plates from the same instrument type, with the same thermal profile and mode, as well as the same block type and sample volume.

Can I run "standard" and "9600 emulation" modes on the 7500 Fast Real-Time PCR System?

Yes, standard and 9600 emulation modes are optionally available. We have matched the ramping rate and thus anticipate equivalent performance. However, the actual total run times will differ from standard and 9600 emulation modes run on a standard block, and we cannot guarantee identical results as performance may vary depending on the assay.

Are there differences in the tube/plate format between Applied Biosystems 7500 Real-Time PCR Systems and Applied Biosystems 7500 Fast Real-Time PCR Systems?

Yes, the standard 7500 instrument has a 96 x 0.2 mL block, whereas the 7500 Fast instrument has a 96 x 0.1 mL block.

Can I import a standard curve for my real-time PCR data into the Design & Analysis Software (v2.x)?

It is possible to import a standard curve into the Design & Analysis Software (v2.x) if you are analyzing results from one of the following Applied Biosystems real-time PCR instruments:
• 7900HT Fast Real-Time PCR System
• 7500/7500 Fast Real-Time PCR System
• StepOne/StepOne Plus Real-Time PCR System
• ViiA 7 Real Time PCR System
• QuantStudio 1 Real-Time PCR System
• QuantStudio 3 & 5 Real-Time PCR System
• QuantStudio 6 & 7 Flex Real-Time PCR System
• QuantStudio 6 & 7 Pro Real-Time PCR Systems
• QuantStudio 12K Flex Real-Time PCR system

Follow the instructions below to import a standard curve for analysis in the Design & Analysis Software (v2.x):
1. Open the data file in the Design & Analysis Software (v2.x).
2. Click on Actions.
3. Click on "Standard Curve Analysis Setting".
4. Select "External Standard Curves" and click on "Import".
5. Browse to the proper .csv file and click "Open".
6. Click "Apply".

What is the difference between a Fast 96-well and standard 96-well block?

There are two main differences between a Fast 96-well and standard 96-well block for the Applied Biosystems real-time PCR systems:
• Reaction well volume: The Fast 96-well blocks have 0.1 mL reaction wells and are compatible with 0.1 mL plates and tubes. The standard 96-well blocks have 0.2 mL reaction wells and are compatible with 0.2 mL plates and tubes.
• Ramp rate: The fast 96-well blocks have a higher maximum ramp rate than standard blocks for the same real-time PCR system. However, both the fast 96-well and standard 96-well blocks are capable of running standard and fast chemistry.

Note: For newer instruments (QuantStudio 3 and 5 Real-Time PCR Systems and later) "0.1 mL" and "0.2 mL" are being used in place of "fast" and "standard" as designations for the two 96-well block formats.

What qPCR instruments are compatible with Thermo Fisher Cloud?

All our real-time PCR instruments are compatible with Thermo Fisher Cloud Apps except the 7500 Fast instrument where the eds files alone are compatible with Thermo Fisher Cloud Apps.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.