Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ 7500 Real Time PCR Systems Spectral Calibration Kit II
GREENER_CHOICE

Catalog No. 4351151
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-511-51 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-511-51 Supplier Applied Biosystems™ Supplier No. 4351151
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This easy-to-use kit enables you to establish the pure dye spectra and multicomponent values for Cy3™, Cy5™, and Texas Red™ dyes on the 7500 Real-Time PCR System.

Applied Biosystems™ 7500 Real-Time PCR Systems Spectral Calibration Kit II contains three optical 96-well reaction plates preloaded and sealed with Cy3™, Cy5™, and Texas Red™ fluorescent dye standards. Spend Your Time on Research, Not Instrument Calibration: This kit is easy-to-use, preloaded, 96-well optical reaction plates enable users to calibrate the 7500 Real-Time PCR System quickly and easily. Insert the reaction plates during system installation, and the system automatically collects the dye standard's spectral information. The system's application algorithm then uses the calibration information to ensure accurate data analysis.

4349180

  • 96-well optical reaction plates preloaded with dyes (FAM™/SYBR™ Green I, VIC™/JOE™, NED™/TAMRA™/ROX™) are provided for quick and easy calibration of the 7500 Real-Time PCR System
  • Easy-to-use and provides accurate discrimination of the fluorescence signal of individual dyes in the reaction mixture
  • Includes the Passive Reference I dye standard for signal normalization in all real-time and endpoint reactions

4351151

  • 96-well optical reaction plates preloaded with Cy3™, Cy5™, and Texas Red™ dyes allow quick and easy calibration of the 7500 Real-Time PCR System
  • Easy-to-use, preloaded spectral calibration kit allows accurate discrimination of the individual dyes in a reaction mixture when all the dyes are excited simultaneously

Spend Your Time on Research, Not Instrument Calibration:
This kit is easy-to-use, preloaded, 96-well optical reaction plates enable users to calibrate the 7500 Real-Time PCR System quickly and easily. Insert the reaction plates during system installation, and the system automatically collects the dye standard's spectral information. The system's application algorithm then uses the calibration information to ensure accurate data analysis.

For Research Use Only. Not for use in diagnostic procedures.

Order Info

Expiration date: 12 months from the date of delivery.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains three Optical 96-Well Reaction Plates, preloaded and sealed containing Cy3™ Texas Red™ and Cy5™ fluorescent dye standards.

Store at -15 to -25°C. (This product is light sensitive.)

Expiration date: 12 months from the date of delivery
Calibrated Dye Cy™3, Cy™5, Texas Red™
For Use With (Equipment) 7500 System
Detection Method Fluorescence
Product Type Spectral Calibration Kit
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Dry Ice
Quantity 1 Kit
Calibration Type Spectral Calibration
What type of Real-Time PCR (Sequence Detection System) information is available from Applied Biosystems?

Tutorials demonstrating introductions to Real-Time PCR, guidelines for designing real-time assays using Primer Express software, and various instrument set-ups can be found under Life Technologies University on our website. User bulletins, manuals and Product inserts, protocols can be found as downloadable pdf files on our Technical Resources page.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.