Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Europium(II) iodide

Catalog No. 7514995G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-149-95G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 75-149-95G Supplier Sigma Aldrich Supplier No. 7514995G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Europium Bromide is a ferromagnetic material that can be used in the synthesis of bromide salt.AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 22015-35-6
Molecular Formula EuI2
Linear Formula EuI2
MDL Number MFCD00799541
Quantity 5 g
Synonym Europium diiodide; Europium iodide
Molecular Weight (g/mol) 405.77
Percent Purity 99.999% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.