Learn More
Enterprise Technology Solutions Paint Marker, Medium Bullet Tip, Black, 6/Pack
SDP
Supplier:  Enterprise Technology Solutions NSN5889099
Encompass_Preferred
ABILITYONE 7520015889099 Skilcraft Pump-action, fiber tip, oil-based paint markers for use on porous and non-porous surfaces including stone, wood, glass, plastic, metal, leather, ceramic, marble, granite, glossy surfaces, paper and more. Ideal for heavy-duty industrial and professional applications as well as for personalization and decorative purposes. Quick-drying paint resists water, sunlight, rough handling and various chemicals. It's weather-proof, fade-proof and acid-free making it archival safe. Ergonomic rubber grip for added comfort and a durable pocket clip on the cap.
Catalog No. 50-204-0664