Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl Methyl Carbonate

Catalog No. 752002100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
50 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-200-2100ML 100 mL
75-200-210ML 10 mL
75-200-250ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 75-200-2100ML Supplier Sigma Aldrich Supplier No. 752002100ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethyl Methyl Carbonate

Specifications

CAS 623-53-0
Density 1.006 g/mL at 25°C
Molecular Formula C4H8O3
Refractive Index n20/D 1.378
Linear Formula C4H8O3
MDL Number MFCD00191398
Quantity 100 mL
Synonym Carbonic acid ethyl methyl ester; EMC; Methyl ethyl carbonate
Molecular Weight (g/mol) 104.1 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.