Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich S-(4-Bromobutyl) thioacetate

Catalog No. 7607141G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-071-41G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 76-071-41G Supplier Sigma Aldrich Supplier No. 7607141G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

S-(4-Bromobutyl) thioacetate is used as a precursor in the formation of organic self-assembled monolayers (SAMs) on metals. Deprotection reactions can then converts the thioacetate group to thiol. These are chemisorbed onto metal surfaces to form self-assembled monolayers with a high degree of structural order. They are often used as a molecular template to form functional nanostructures.Used in the formation of Self-Assembled Monolayers on gold electrodes to modify its work function.

Specifications

CAS 14475-59-3
Density 1.388 g/mL (at 25°C)
Molecular Formula C6H11BrOS
Linear Formula C6H11BrOS
Refractive Index n20/D 1.518
MDL Number MFCD22666439
Quantity 1 g
Synonym 1-Butanethiol, 4-bromo-, acetate; Acetic acid, thio-,S-(4-bromobutyl) ester; Ethanethioic acid, S-(4-bromobutyl) ester
Molecular Weight (g/mol) 211.12
Percent Purity 96% (GC)
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.