Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dibenzocyclooctyne-Sulfo-N-Hydroxysuccinimidyl Ester

Catalog No. 76204050MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-204-050MG 50 mg
76-204-01MG 1 mg
76-204-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 76-204-050MG Supplier Sigma Aldrich Supplier No. 76204050MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dibenzocyclooctyne-Sulfo-N-Hydroxysuccinimidyl Ester

Specifications

Melting Point 240°C to 260°C
Molecular Formula C25H21N2NaO8S
Linear Formula C25H21N2NaO8S
MDL Number MFCD22572669
Quantity 50 mg
Synonym DBCO-sulfo-NHS ester; DBCO-sulfo-SE
Molecular Weight (g/mol) 532.5 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.