Learn More
SHOWA™ 7710 Series PVC Coated General Purpose Gloves, Cotton Lining
Catalog No. 19019325 Shop All Showa Best Glove Products
Change view
Click to view available options
Grip Design:
Rough
Smooth
Series:
7710 Series
7710R Series
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grip Design Series
19-019-325 Rough 7710R Series
11-395-14 Smooth 7710 Series
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-019-325 Supplier SHOWA™ Supplier No. 7710R10
This item is not returnable. View return policy
Multi-purpose gloves with cotton lining provides defensive protection from chemicals, oil and grease.
Both styles are ideal for working with chemicals, oil, or grease in refining, mining, automotive subassembly, or construction
  • Fully coated PVC with smooth or rough finish
  • Cotton jersey lined Sanitized™
  • Recyclable
  • Washable
Specifications
Certifications/Compliance CFIA
Color Black
Latex Free Yes
Material PVC
Size Large
For Use With (Application) Chemical spray and treatment, coating preparation, laboratory, pharma and analysis, painting and spray, washing and cleaning
Glove Cuff Style Gauntlet
Grip Design Rough
Liner Material Cotton Jersey
Lining Lined
Model 7710R-10
Series 7710R Series
Length (English) 10 in.
Length (Metric) 25.4 cm
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.