Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ 7712 Series PVC Coated Gloves

Catalog No. 19019327 Shop All Showa Best Glove Products
Change view
Click to view available options
Grip Design:
Rough
Smooth
Series:
7712 Series
7712R Series
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grip Design Series
19-019-327 Smooth 7712 Series
11-395-15 Rough 7712R Series
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-019-327 Supplier SHOWA™ Supplier No. 771210
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PVC coated, multi-purpose gloves with soft cotton liner are designed for all industrial applications and protect against a range of hazards including chemicals, oils and abrasion.

Both styles are ideal for working with chemicals, oil, or grease in refining, mining, automotive subassembly, or construction

  • Fully coated PVC with smooth or rough finish
  • Cotton jersey lined Sanitized™
  • Recyclable
  • Washable

Specifications

Certifications/Compliance CFIA
Color Black
Latex Free Yes
Material PVC
Size Large
For Use With (Application) Chemical spray and treatment, coating preparation, laboratory, pharma and analysis, washing and cleaning
Glove Cuff Style Gauntlet
Grip Design Smooth
Liner Material Cotton
Lining Lined
Model 7712-10
Series 7712 Series
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.