Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pentafluoropropionic anhydride

Catalog No. 7729225ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-292-25ML 25 mL
77-292-5ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 77-292-25ML Supplier Sigma Aldrich Supplier No. 7729225ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Pentafluoropropionic anhydride can be used to synthesize lithium 4,5-dicyano-2-(pentafluoroethyl)imidazole, a novel imidazole-derived salt with potential application as an electrolyte in lithium-ion batteries
  • It can also be used to prepare 1.3- bis(pentafluoropropionyl)azulene

Specifications

CAS 356-42-3
Density 1.571 g/mL (at 25°C (literature))
Boiling Point 69°C to 70°C (735 mmHg)
Molecular Formula C6F10O3
Linear Formula (CF3CF2CO)2 O
MDL Number MFCD00000429
Quantity 25 mL
Synonym PFPA; Perfluoropropionic anhydride
Molecular Weight (g/mol) 310.05
Percent Purity ≥97.0% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.