Learn More
Sigma Organic Chemistry Copper | 5G | 7440-50-8 | MFCD00010965 | >=99.5% trace metals basis
Supplier: Sigma Organic Chemistry 7741115G
Copper, 5G
About This Item:
Linear Formula: Cu
Storage: room temp
EINECS Number: 231-159-6
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.