Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 12-Pentafluorophenoxydodecylphosphonic acid

Catalog No. 777188500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-718-8500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 77-718-8500MG Supplier Sigma Aldrich Supplier No. 777188500MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

12-Pentafluorophenoxydodecylphosphonic acid

Specifications

CAS 1049677-16-8
Melting Point 80°C to 85°C
Molecular Formula C18H26F5O4P
Linear Formula C18H26F5O4P
Quantity 500 mg
MDL Number MFCD11046096
Synonym P-[12-(2,3,4,5,6-Pentafluorophenoxy)dodecyl]phosphonic acid
Molecular Weight (g/mol) 432.36
Percent Purity 99% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.