Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Butyl-3-Methylimidazolium Bis(Trifluoromethylsulfonyl)Imide

Catalog No. 778961GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-896-1GF 10 kg
77-896-5GF 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 77-896-1GF Supplier Sigma Aldrich Supplier No. 778961GF
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Butyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide is a room temperature ionic liquid (RTIL). The solubility of alkane gases are less than the alkene gas in this IL. Its ability to solubilize CO2 is more when compared to 1-butyl-3-methylimidazolium dicyanamide.Product protected by U.S. Patent 5,827,602 assigned to Covalent Associates, Inc. Woburn, MA, USA

Specifications

CAS 128446-35-5
Refractive Index n20/D 1.428
Quantity 10 kg
Synonym BMIIm; BMIM TFSI
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.