Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Gamma-Cyclodextrin
Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
100 g
Description
+a-Cyclodextrin (+a-CD), a water-soluble cyclic oligosaccharide consisting of eight +a-(1,4)-linked glucopyranose subunits, can be used in a wide range of applications including:Synthesis of renewable and biocompatible metal-organic frameworks.Formation of inclusion complexes with a variety of guest molecules to increase their solubility in water and other polar solvents.Synthesis of +a-CD based nanogels and dendrimers as carriers and stabilizers for drug delivery applications.Cavamax is a registered trademark of Wacker Chemie AG
Specifications
Specifications
| CAS | 128446-36-6 |
| Melting Point | 180°C to 182°C |
| Quantity | 100 g |
| Synonym | Cavasol(R) W7 M; Methyl-beta-cyclodextrin |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction