Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetrabutylammonium Hydrogensulfate

Catalog No. 791784500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-178-4500G 100 g
79-178-41KG 500 g
79-178-4100G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 79-178-4500G Supplier Sigma Aldrich Supplier No. 791784500G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetrabutylammonium Hydrogensulfate

Specifications

CAS 32503-27-8
Melting Point 169°C to 171°C
Molecular Formula C16H37NO4S
Linear Formula (CH3CH2CH2CH2)4N(HSO4)
MDL Number MFCD00011637
Quantity 100 g
Synonym Tetrabutylammonium bisulfate
Molecular Weight (g/mol) 339.53 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.