Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phosphazene Base P2-Et

Catalog No. 794171ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-417-1ML 250 mg
79-417-5ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 79-417-1ML Supplier Sigma Aldrich Supplier No. 794171ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phosphazene base P2-Et is an extremely strong, neutral nitrogen base. It gives excellent results where other bases failThis strong non-ionic base has the ability to catalyze a wide variety of organic transformations such as:Palladium-catalyzed cross-coupling reactions when used in combination with tBuXPhos or tBuBrettPhos G3 precatalysts.Deprotonation of ortho-halobenzyl sulfones to generate +a-sulfonyl benzylic carbanions.Conversion of vinyl sufone to allyl sulfone.+a-Alkylation of 2-phenyl-2-oxazoline-4-carbonylcamphorsultam in the presence of tetrabutylammonium bromide.

Specifications

CAS 1442433-71-7
Molecular Formula C23H22BF4N
Linear Formula C23H22BF4N
Refractive Index n20/D 1.492; n20/D 1.492
Quantity 250 mg
Synonym 9-Mesityl-10-methylacridinium tetrafluoroborate; 10-Methyl-9-(2,4,6-trimethylphenyl)acridinium tetrafluoroborate
Molecular Weight (g/mol) 399.23 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.