Learn More
Abcam 7alpha-Hydroxycholesterol, Bile Acid Precursor, 1 mg
Description
- Molecular Weight: 402.65 g/mol
- Biological Function: Bile acid precursor produced by cholesterol 7 alpha-hydroxylase. Induces inflammation by enhancing production of chemokine (C-C motif) ligand 2 and elicits TLR6-mediated expression of IL-23 in monocytic cells.
- Composition: 100% 7α-Hydroxycholesterol
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Target Molecule | 7α-Hydroxycholesterol |
| Content And Storage | Store at -20°C. |
| Buffer | 100% 7α-Hydroxycholesterol |
| Shipping Condition | Ambient |
| Formulation | 100% 7α-Hydroxycholesterol |
| Quantity | 1 mg |
| For Use With (Application) | Sample preparation, detection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.