Learn More
Leica Biosystems 7ml AFP Bond RTU Primary
Supplier:  Leica Biosystems PA0963U
Encompass
Antigen BackgroundAlpha Fetoprotein (AFP) is an oncofetal antigen of 7 kD found in body fluids, which if detected in high concentrations has clinical implications. AFP is expressed in fetal liver but is not present under normal circumstances in healthy adult tissues. It is reported to be expressed in a proportion of germ cell tumors, with high frequency in yolk sac tumors.DisclaimerAlpha Fetoprotein is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-2015