Learn More
Leica Biosystems 7ml Bcl-2 Bond RTU Primary
Supplier:  Leica Biosystems PA0117U
Encompass
Antigen BackgroundBcl-2 is a member of a family of proteins that are involved in apoptosis. Bcl-2 is an integral inner mitochondrial membrane protein of 25 kD and has a wide tissue distribution. It is considered to act as an inhibitor of apoptosis. For this reason, bcl-2 expression is inhibited in germinal centers where apoptosis forms part of the B cell production pathway.In 9% of follicular lymphomas a translocation occurs which juxtaposes the bcl-2 gene at 18q21, to an immunoglobulin gene. This t(14;18) translocation can deregulate gene expression and bcl-2 over-expression can be demonstrated immunohistochemically in the vast majority of follicular lymphomas.DisclaimerBcl-2 Oncoprotein is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-2156
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.