Learn More
Leica Biosystems 7ml CD15 (MMA) Bond RTU Primar
Supplier:  Leica Biosystems PA0473U
Encompass
Antigen BackgroundCD15 antigen, also known as X-hapten, is reported to be expressed on 9% of circulating human granulocytes, 3-6% of circulating monocytes and is absent from normal lymphocytes. The CD15 antigen is also expressed on Reed Sternberg cells of Hodgkin's disease and some leukemias.DisclaimerCD15 is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-2078
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.