Learn More
Leica Biosystems 7ml CD23 Bond RTU Primary
Supplier:  Leica Biosystems PA0169U
Encompass
Antigen BackgroundThe CD23 molecule is the low affinity IgE receptor found on B cells.It is a membrane glycoprotein of 45 kD and is reported to be found on a sub-population of peripheral blood cells, B lymphocytes and on EBV-transformed B lymphoblastoid cell lines.Expression of CD23 antigen has been reported on monocytes and dendritic cells.DisclaimerCD23 is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-2142
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.