Learn More
Leica Biosystems 7ml CD56 Bond RTU Primary
Supplier:  Leica Biosystems PA0191U
Encompass
Antigen BackgroundThe neural cell adhesion molecules are a family of closely-related cell surface glycoproteins thought to play a role in embryogenesis, development and contact-mediated interactions between neural cells. The CD56 antigen (NCAM) consists of four major isoforms generated by differential splicing of the RNA transcript from a single gene located on chromosome 5. The CD56 antigen is expressed on neurons, astrocytes, Schwann cells, NK cells and a subset of activated T lymphocytes.DisclaimerCD56 (NCAM) is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-2138
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.