Learn More
Leica Biosystems 7ml Cytokeratin 20 Bond RTU Pr
Supplier:  Leica Biosystems PA0022U
Encompass
Antigen BackgroundCytokeratin 2 has been demonstrated to be almost entirely confined to the gastric and intestinal epithelium, urothelium and Merkel cells of the skin. Cytokeratin 2 is less acidic than other type I cytokeratins and is of interest due to its restricted tissue expression. In normal tissue, cytokeratin 2 is expressed in intestinal epithelium, gastric foveolar epithelium, a number of endocrine cells in the upper portions of the pyloric glands, urothelium and Merkel cells in epidermis. In tumors it is reported, there is a marked difference in the expression of cytokeratin 2 within different carcinomas. Neoplasms expressing cytokeratin 2 are derived from normal epithelia which themselves expressed cytokeratin 2. Colorectal carcinomas consistently express cytokeratin 2, while gastric adenocarcinomas express cytokeratin 2 to a lesser degree. Adenocarcinomas of the gall bladder and bile duct, ductal cell adenocarcinomas of the pancreas, mucinous ovarian tumors, Merkel cell tu
Catalog No. 50-255-2182
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.