Learn More
Leica Biosystems 7ML Ep-CAM (MOC31 Mab) 'CM'
Supplier:  Leica Biosystems PA0797
Encompass
This antibody is intended for in vitro diagnostic (IVD) use. The Cell Marque Ep-CAM (MOC-31) antibody is intended for qualified laboratories to qualitatively identify by light microscopy the presence of associated antigens in sections of formalin-fixed, paraffin-embedded tissue sections using IHC test methods. Use of this antibody is indicated, subsequent to clinical differential diagnosis, as an aid in the identification of epithelial cells as well as adenocarcinoma versus maligant mesothelioma and basal cell carcinoma versus squamous cell carcinoma of the skin within the context of an antibody panel, the patient’s clinical history, and other diagnostic tests evaluated by a qualified pathologist.Instructions For Use (IFU)Leica Biosystems has not validated the performance characteristics of the Cell Marque products on the BOND and BOND-MAX analyzers. For performance data, MSDS sheets and technical trouble-shooting please contact Cell Marque directly.
Catalog No. 50-255-2043
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.