Learn More
Leica Biosystems 7ml HGH Bond RTU Primary
Supplier:  Leica Biosystems PA0704U
Encompass
Antigen BackgroundGrowth hormone (GH), somatotropin, is the primary hormone responsible for regulating overall body growth and is also important in organic metabolism. It is synthesized by acidophilic or somatotropic cells of the anterior pituitary gland. Human GH has a molecular weight of 22 kD.GH stimulates growth indirectly by promoting the liver's production of somatomedins, which act directly on bone and soft tissue to cause growth. GH exerts direct metabolic effects on the liver, adipose tissue and muscle. In general, growth hormone enhances protein synthesis, conserves carbohydrates and uses up fat stores.DisclaimerHuman Growth Hormone (HGH) is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-2056
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.