Learn More
Leica Biosystems 7ml MSH2 (79H11) Bond RTU Prim
Supplier:  Leica Biosystems PA0989U
Encompass
Antigen BackgroundHuman mismatch repair protein 2 (MSH2) is involved in the initial recognition of mismatched nucleotides during the post replication mismatch repair process. Therefore, the loss of MSH2 function leads to the accumulation of replication errors, which in turn may be responsible for the multiple mutations required for multistage carcinogenesis. MSH2 is reported to be expressed in the nuclei of cells from a variety of tissues including thyroid, heart, smooth muscle and the germinal centers of lymphoid follicles. In ileum and colon, MSH2 expression has been reported in the crypts, the cells which are undergoing rapid renewal. They are responsible for the continuous production of differentiated cells which migrate over 2 to 4 days before being sloughed into the lumen.DisclaimerMismatch Repair Protein (MSH2) is a recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunol
Catalog No. 50-255-2012
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.