Learn More
Leica Biosystems 7ml NSE Bond RTU Primary
Supplier:  Leica Biosystems PA0435U
Encompass
Antigen BackgroundClone 22C9 reacts with the gamma subunit of the enolase isoenzyme.DisclaimerNeuron Specific Enolase is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-2085
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.